missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Methoxyphenyl)piperazine 98.0+%, TCI America™
$61.51 - $156.59
Chemical Identifiers
| CAS | 35386-24-4 |
|---|---|
| Molecular Formula | C11H18Cl2N2O |
| Molecular Weight (g/mol) | 265.18 |
| MDL Number | MFCD00005958 |
| InChI Key | ZGWQDMTYAQEMHA-UHFFFAOYSA-N |
| Synonym | 1-2-methoxyphenyl piperazine, 1-2-methoxy-phenyl-piperazine, 1-o-methoxyphenyl piperazine, 2-mpp, 1-2-methoxyphenyl-piperazine, n-2-methoxyphenyl piperazine, unii-81njo1330a, 1-2-methoxy phenyl piperazine, 2-methoxyphenylpiperazine, piperazine, 1-2-methoxyphenyl |
| PubChem CID | 1346 |
| IUPAC Name | dihydrogen 1-(2-methoxyphenyl)piperazine dichloride |
| SMILES | [H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1N1CCNCC1 |
Chemical Identifiers
| 35386-24-4 | |
| 265.18 | |
| ZGWQDMTYAQEMHA-UHFFFAOYSA-N | |
| 1346 | |
| [H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1N1CCNCC1 |
| C11H18Cl2N2O | |
| MFCD00005958 | |
| 1-2-methoxyphenyl piperazine, 1-2-methoxy-phenyl-piperazine, 1-o-methoxyphenyl piperazine, 2-mpp, 1-2-methoxyphenyl-piperazine, n-2-methoxyphenyl piperazine, unii-81njo1330a, 1-2-methoxy phenyl piperazine, 2-methoxyphenylpiperazine, piperazine, 1-2-methoxyphenyl | |
| dihydrogen 1-(2-methoxyphenyl)piperazine dichloride |
Specifications
| 35386-24-4 | |
| MFCD00005958 | |
| 3263 | |
| ZGWQDMTYAQEMHA-UHFFFAOYSA-N | |
| dihydrogen 1-(2-methoxyphenyl)piperazine dichloride | |
| 1346 | |
| ≥98.0% (GC,T) | |
| 1-(2-Methoxyphenyl)piperazine |
| C11H18Cl2N2O | |
| 5 g | |
| 1-2-methoxyphenyl piperazine, 1-2-methoxy-phenyl-piperazine, 1-o-methoxyphenyl piperazine, 2-mpp, 1-2-methoxyphenyl-piperazine, n-2-methoxyphenyl piperazine, unii-81njo1330a, 1-2-methoxy phenyl piperazine, 2-methoxyphenylpiperazine, piperazine, 1-2-methoxyphenyl | |
| [H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1N1CCNCC1 | |
| 265.18 | |
| 192.26 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No