missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methoxyphenyl)piperazine 98.0+%, TCI America™

Catalog No. M08835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08835G 5 g
M088325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08835G Supplier TCI America Supplier No. M08835G
Only null left

Chemical Identifiers

CAS 35386-24-4
Molecular Formula C11H18Cl2N2O
Molecular Weight (g/mol) 265.18
MDL Number MFCD00005958
InChI Key ZGWQDMTYAQEMHA-UHFFFAOYSA-N
Synonym 1-2-methoxyphenyl piperazine, 1-2-methoxy-phenyl-piperazine, 1-o-methoxyphenyl piperazine, 2-mpp, 1-2-methoxyphenyl-piperazine, n-2-methoxyphenyl piperazine, unii-81njo1330a, 1-2-methoxy phenyl piperazine, 2-methoxyphenylpiperazine, piperazine, 1-2-methoxyphenyl
PubChem CID 1346
IUPAC Name dihydrogen 1-(2-methoxyphenyl)piperazine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1N1CCNCC1

Specifications

Quantity 5 g
UN Number 3263
Formula Weight 192.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-(2-Methoxyphenyl)piperazine