missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methoxyethyl)piperazine 98.0+%, TCI America™

Catalog No. M21691G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21691G 1 g
M21695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21691G Supplier TCI America Supplier No. M21691G

Chemical Identifiers

CAS 13484-40-7
Molecular Formula C7H16N2O
Molecular Weight (g/mol) 144.218
MDL Number MFCD00191214
InChI Key BMEMBBFDTYHTLH-UHFFFAOYSA-N
Synonym 1-2-methoxyethyl piperazine, 1-2-methoxyethyl-piperazine, 1-2-methoxy-ethyl-piperazine, piperazine, 1-2-methoxyethyl, 1-methoxy-2-piperazinylethane, pubchem14209, 2-methoxyethylpiperazine, n-methoxyethyl piperazine, 2-methoxyethyl piperazine, n-methoxyethylylpiperazine
PubChem CID 2734638
IUPAC Name 1-(2-methoxyethyl)piperazine
SMILES COCCN1CCNCC1

Specifications

Color Yellow
Boiling Point 101°C
Quantity 1 g
Formula Weight 144.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(2-Methoxyethyl)piperazine