missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methoxyethyl)-1-methylpyrrolidinium Bis(fluorosulfonyl)imide 98.0+%, TCI America™

Catalog No. M29985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29985G 5 g
M29981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29985G Supplier TCI America Supplier No. M29985G

Chemical Identifiers

CAS 1235234-47-5
Molecular Formula C8H18F2N2O5S2
Molecular Weight (g/mol) 324.358
InChI Key HBTKRUXLKWRXSK-UHFFFAOYSA-N
PubChem CID 132274865
IUPAC Name bis(fluorosulfonyl)azanide;1-(2-methoxyethyl)-1-methylpyrrolidin-1-ium
SMILES C[N+]1(CCCC1)CCOC.[N-](S(=O)(=O)F)S(=O)(=O)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 324.36
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-(2-Methoxyethyl)-1-methylpyrrolidinium Bis(fluorosulfonyl)imide