missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyethyl)pyrrolidine 98.0+%, TCI America™

Catalog No. p-7130557
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H047925ML 25 mL
H0479100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H047925ML Supplier TCI America Supplier No. H047925ML

Chemical Identifiers

CAS 2955-88-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00003181
InChI Key XBRDBODLCHKXHI-UHFFFAOYSA-N
Synonym 1-pyrrolidineethanol, n-2-hydroxyethyl pyrrolidine, 1-2-hydroxyethyl pyrrolidine, 2-pyrrolidin-1-yl ethanol, epolamine, 2-pyrrolidinoethanol, pyrrolidinoethanol, hydroxyethylpyrrolidine, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol
PubChem CID 76288
ChEBI CHEBI:48293
IUPAC Name 2-(pyrrolidin-1-yl)ethan-1-ol
SMILES OCCN1CCCC1

Specifications

Color Yellow
Quantity 25 mL
UN Number 1993
Formula Weight 115.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(2-Hydroxyethyl)pyrrolidine