missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyethyl)pyrrolidine 98.0+%, TCI America™

Catalog No. h047925ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
Catalog No. Quantity
H047925ML 25 mL
H0479100ML 100 mL
2 options

Catalog No. H047925ML

Supplier: TCI America H047925ML

Only null left

Chemical Identifiers

CAS 2955-88-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00003181
InChI Key XBRDBODLCHKXHI-UHFFFAOYSA-N
Synonym 1-pyrrolidineethanol, n-2-hydroxyethyl pyrrolidine, 1-2-hydroxyethyl pyrrolidine, 2-pyrrolidin-1-yl ethanol, epolamine, 2-pyrrolidinoethanol, pyrrolidinoethanol, hydroxyethylpyrrolidine, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol
PubChem CID 76288
ChEBI CHEBI:48293
IUPAC Name 2-(pyrrolidin-1-yl)ethan-1-ol
SMILES OCCN1CCCC1

Specifications

Color Yellow
Quantity 25 mL
UN Number 1993
Formula Weight 115.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(2-Hydroxyethyl)pyrrolidine