missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1-(2-Hydroxyethyl)piperidine

Catalog No. 116068500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
116068500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 116068500G Supplier Sigma Aldrich Supplier No. 116068500G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Counter-ion used for investigations of skin permeation of flurbiprofencatalyst used for enantioselective synthesisamine quencher used during synthesis of photoacid generators for arf lithographyadsorbent for capture and release of pressurized carbon dioxide for greenhouse gas controlreagentplus is a registered trademark of sigma-aldrich co
  • Llc

Specifications

CAS 3040-44-6
Density 0.973 g/mL (at 25°C (literature))
Boiling Point 199°C to 202°C (lit.)
Molecular Formula C7H15NO
Refractive Index n20/D 1.4804 (literature)
Linear Formula C7H15NO
Quantity 500 g
MDL Number MFCD00006512
Synonym 1-Piperidineethanol; 2-Piperidinoethanol
Molecular Weight (g/mol) 129.2
Percent Purity 99%
Show More Show Less