missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyethyl)piperazine 99.0+%, TCI America™

Catalog No. P0812500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0812500G 500 g
P081225G 25 g
P0812100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0812500G Supplier TCI America Supplier No. P0812500G

Chemical Identifiers

CAS 103-76-4
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.191
MDL Number MFCD00005970
InChI Key WFCSWCVEJLETKA-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl piperazine, 1-piperazineethanol, 1-2-hydroxyethyl piperazine, hydroxyethylpiperazine, 2-1-piperazinyl ethanol, 2-hydroxyethylpiperazine, 2-piperazinoethanol, 2-piperazin-1-yl ethan-1-ol, 1-piperazinethanol, 4-2-hydroxyethyl piperazine
PubChem CID 7677
IUPAC Name 2-piperazin-1-ylethanol
SMILES C1CN(CCN1)CCO

Specifications

Color Colorless
Boiling Point 246°C
Quantity 500 g
Formula Weight 130.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Hydroxyethyl)piperazine
EINECSNumber (5)-0957&(5)-3833
RTECSNumber TL6825000
TSCA Yes