Learn More
1-(2-Hydroxyethyl)piperazine, 98+%
CAS: 103-76-4 | C6H14N2O | 130.191 g/mol
Supplier: Thermo Scientific Chemicals A1721522
Description
1-Piperazineethanol is a hydroxyalkyl substituted pieprazine used in the preparation of pharmaceutical compounds. It is also used as an Intermediate in the manufacture of polyurethane catalysts, corrosion inhibitors, surfactants, synthetic fibers, pharmaceuticals. It is also used to synthesize triethylenediamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Piperazineethanol is a hydroxyalkyl substituted pieprazine used in the preparation of pharmaceutical compounds. It is also used as an Intermediate in the manufacture of polyurethane catalysts, corrosion inhibitors, surfactants, synthetic fibers, pharmaceuticals. It is also used to synthesize triethylenediamine.
Solubility
Miscible with water. Also miscible in ethanol, acetone, ether benzene. Immiscible in hexane.
Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place.
Specifications
| 1-(2-Hydroxyethyl)piperazine | |
| -39°C | |
| 12.3 | |
| 135°C (275°F) | |
| C6H14N2O | |
| MFCD00005970 | |
| 104361 | |
| Miscible with water. Also miscible in ethanol,acetone,ether benzene. Immiscible in hexane. | |
| C1CN(CCN1)CCO | |
| 130.191 | |
| 130.19 |
| 103-76-4 | |
| 1.06 | |
| 242°C to 246°C | |
| Amine-like | |
| 1.5065 | |
| 100 g | |
| n-2-hydroxyethyl piperazine, 1-piperazineethanol, 1-2-hydroxyethyl piperazine, hydroxyethylpiperazine, 2-1-piperazinyl ethanol, 2-hydroxyethylpiperazine, 2-piperazinoethanol, 2-piperazin-1-yl ethan-1-ol, 1-piperazinethanol, 4-2-hydroxyethyl piperazine | |
| WFCSWCVEJLETKA-UHFFFAOYSA-N | |
| 2-piperazin-1-ylethanol | |
| 7677 | |
| ≥98% |
Chemical Identifiers
| 103-76-4 | |
| 130.191 | |
| WFCSWCVEJLETKA-UHFFFAOYSA-N | |
| 7677 | |
| C1CN(CCN1)CCO |
| C6H14N2O | |
| MFCD00005970 | |
| n-2-hydroxyethyl piperazine, 1-piperazineethanol, 1-2-hydroxyethyl piperazine, hydroxyethylpiperazine, 2-1-piperazinyl ethanol, 2-hydroxyethylpiperazine, 2-piperazinoethanol, 2-piperazin-1-yl ethan-1-ol, 1-piperazinethanol, 4-2-hydroxyethyl piperazine | |
| 2-piperazin-1-ylethanol |
Safety and Handling
GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.
P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
H315-H318
EINECSNumber : 203-142-3
RTECSNumber : TL6825000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only