missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyethyl)piperazine, 98+%

Catalog No. AAA1721509
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1721509 10 g
AAA1721522 100 g
AAA1721536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1721509 Supplier Thermo Scientific Chemicals Supplier No. A1721509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103-76-4 | C6H14N2O | 130.191 g/mol

1-Piperazineethanol is a hydroxyalkyl substituted pieprazine used in the preparation of pharmaceutical compounds. It is also used as an Intermediate in the manufacture of polyurethane catalysts, corrosion inhibitors, surfactants, synthetic fibers, pharmaceuticals. It is also used to synthesize triethylenediamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Piperazineethanol is a hydroxyalkyl substituted pieprazine used in the preparation of pharmaceutical compounds. It is also used as an Intermediate in the manufacture of polyurethane catalysts, corrosion inhibitors, surfactants, synthetic fibers, pharmaceuticals. It is also used to synthesize triethylenediamine.

Solubility
Miscible with water. Also miscible in ethanol, acetone, ether benzene. Immiscible in hexane.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 103-76-4
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.191
MDL Number MFCD00005970
InChI Key WFCSWCVEJLETKA-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl piperazine, 1-piperazineethanol, 1-2-hydroxyethyl piperazine, hydroxyethylpiperazine, 2-1-piperazinyl ethanol, 2-hydroxyethylpiperazine, 2-piperazinoethanol, 2-piperazin-1-yl ethan-1-ol, 1-piperazinethanol, 4-2-hydroxyethyl piperazine
PubChem CID 7677
IUPAC Name 2-piperazin-1-ylethanol
SMILES C1CN(CCN1)CCO

Specifications

Melting Point -39°C
Density 1.06
pH 12.3
Boiling Point 242°C to 246°C
Flash Point 135°C (275°F)
Odor Amine-like
Refractive Index 1.5065
Quantity 10 g
Beilstein 104361
Solubility Information Miscible with water. Also miscible in ethanol,acetone,ether benzene. Immiscible in hexane.
Formula Weight 130.19
Percent Purity ≥98%
Chemical Name or Material 1-(2-Hydroxyethyl)piperazine
Show More Show Less
Hazard Category H315-H318
Hazard Statement GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
EINECSNumber 203-142-3
RTECSNumber TL6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only