missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyethyl)-4-methylpiperazine, 98%, Thermo Scientific™

Chemical Identifiers

CAS 5464-12-0
Molecular Formula C7H16N2O
Molecular Weight (g/mol) 144.22
MDL Number MFCD03840691
InChI Key QHTUMQYGZQYEOZ-UHFFFAOYSA-N
Synonym 1-2-hydroxyethyl-4-methylpiperazine, 2-4-methylpiperazin-1-yl ethanol, 1-piperazineethanol, 4-methyl, 2-4-methylpiperazin-1-yl ethan-1-ol, 1-n-hydroxyethyl-4-methyl piperazine, 2-4-methyl-piperazin-1-yl-ethanol, 1-2-hydroxyethyl-4-methyl-piperazine, 4-methylpiperazine-1-ethanol, 2-4-methylpiperazinyl ethan-1-ol, 2-4-methyl-1-piperazinyl ethanol
PubChem CID 231184
IUPAC Name 2-(4-methylpiperazin-1-yl)ethanol
SMILES CN1CCN(CC1)CCO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC384832500
SDS View Documents
Thermo Scientific Chemicals
384832500
250mg Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC384830010
SDS View Documents
Thermo Scientific Chemicals
384830010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC384830050
SDS View Documents
Thermo Scientific Chemicals
384830050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5464-12-0
144.22
QHTUMQYGZQYEOZ-UHFFFAOYSA-N
231184
CN1CCN(CC1)CCO
C7H16N2O
MFCD03840691
1-2-hydroxyethyl-4-methylpiperazine, 2-4-methylpiperazin-1-yl ethanol, 1-piperazineethanol, 4-methyl, 2-4-methylpiperazin-1-yl ethan-1-ol, 1-n-hydroxyethyl-4-methyl piperazine, 2-4-methyl-piperazin-1-yl-ethanol, 1-2-hydroxyethyl-4-methyl-piperazine, 4-methylpiperazine-1-ethanol, 2-4-methylpiperazinyl ethan-1-ol, 2-4-methyl-1-piperazinyl ethanol
2-(4-methylpiperazin-1-yl)ethanol

Specifications

5464-12-0
100.0
Authentic
Glass Bottle
1.4850 to 1.487
250mg
QHTUMQYGZQYEOZ-UHFFFAOYSA-N
2-(4-methylpiperazin-1-yl)ethanol
231184
98%
97.5
54°C to 55°C (0.5 mbar)
97.5% min. (GC)
C7H16N2O
MFCD03840691
1-2-hydroxyethyl-4-methylpiperazine, 2-4-methylpiperazin-1-yl ethanol, 1-piperazineethanol, 4-methyl, 2-4-methylpiperazin-1-yl ethan-1-ol, 1-n-hydroxyethyl-4-methyl piperazine, 2-4-methyl-piperazin-1-yl-ethanol, 1-2-hydroxyethyl-4-methyl-piperazine, 4-methylpiperazine-1-ethanol, 2-4-methylpiperazinyl ethan-1-ol, 2-4-methyl-1-piperazinyl ethanol
CN1CCN(CC1)CCO
144.22
144.22
1-(2-Hydroxyethyl)-4-methylpiperazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only