missing translation for 'onlineSavingsMsg'
Learn More

1,2-Hexadecanediol 98.0+%, TCI America™

$136.08 - $404.51

Chemical Identifiers

CAS 6920-24-7
Molecular Formula C16H34O2
Molecular Weight (g/mol) 258.45
MDL Number MFCD00004727
InChI Key BTOOAFQCTJZDRC-UHFFFAOYNA-N
Synonym 1,2-Dihydroxyhexadecane
PubChem CID 98037
ChEBI CHEBI:75586
IUPAC Name hexadecane-1,2-diol
SMILES CCCCCCCCCCCCCCC(O)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H09935G
View Documents
TCI America
H09935G
5 g N/A
Only null left
 
H099325G
View Documents
TCI America
H099325G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6920-24-7
258.45
BTOOAFQCTJZDRC-UHFFFAOYNA-N
98037
hexadecane-1,2-diol
C16H34O2
MFCD00004727
1,2-Dihydroxyhexadecane
CHEBI:75586
CCCCCCCCCCCCCCC(O)CO

Specifications

6920-24-7
White
MFCD00004727
1,2-Dihydroxyhexadecane
CCCCCCCCCCCCCCC(O)CO
258.45
CHEBI:75586
≥98.0% (GC)
1,2-Hexadecanediol
76°C
C16H34O2
5 g
BTOOAFQCTJZDRC-UHFFFAOYNA-N
hexadecane-1,2-diol
98037
258.45
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'