missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Hexadecanediol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 76°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 258.45 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,2-Hexadecanediol |
Safety and Handling
| TSCA | Yes |