missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Furoyl)piperazine Hydrochloride 98.0+%, TCI America™

Catalog No. F07055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07055G 5 g
F070525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07055G Supplier TCI America Supplier No. F07055G

Chemical Identifiers

CAS 60548-09-6
Molecular Formula C9H13ClN2O2
Molecular Weight (g/mol) 216.67
MDL Number MFCD00236012
InChI Key FMFUHCXDFVDINI-UHFFFAOYSA-N
Synonym 1-[(2-Furan)carbonyl]piperazine Hydrochloride
PubChem CID 12289910
IUPAC Name 1-(furan-2-carbonyl)piperazine hydrochloride
SMILES Cl.O=C(N1CCNCC1)C1=CC=CO1

Specifications

Melting Point 207°C
Quantity 5 g
Formula Weight 216.67
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Furoyl)piperazine Hydrochloride