missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluorophenyl)piperazine Hydrochloride 98.0+%, TCI America™

Catalog No. F02915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02915G 5 g
F029125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02915G Supplier TCI America Supplier No. F02915G
Only null left

Chemical Identifiers

CAS 1011-16-1
Molecular Formula C10H14ClFN2
Molecular Weight (g/mol) 216.684
MDL Number MFCD00134409
InChI Key JVQOFQHGEFLRRQ-UHFFFAOYSA-N
Synonym 1-2-fluorophenyl piperazine hydrochloride, n-2-fluorophenyl piperazine hydrochloride, 1-2-fluorophenyl piperazine monohydrochloride, 1-2-fluorophenyl piperazine hcl, pubchem15282, timtec-bb sbb003382, acmc-2097ud, labotest-bb lt00160127, ksc495m4b, 2-fluorophenyl piperazine, chloride
PubChem CID 2734698
IUPAC Name 1-(2-fluorophenyl)piperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC=CC=C2F.Cl

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 5 g
Formula Weight 216.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Fluorophenyl)piperazine Hydrochloride