missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluoro-4-nitrophenyl)homopiperazine, 97%, Thermo Scientific Chemicals

Catalog No. AAH6331006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6331006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6331006 Supplier Thermo Scientific Chemicals Supplier No. H6331006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 392660-27-4 | Molecular Formula: C11H14FN3O2 | Molecular Weight: 239.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 392660-27-4
Molecular Formula C11H14FN3O2
Molecular Weight (g/mol) 239.25
InChI Key BSSBZGVNKLRQEL-UHFFFAOYSA-N
Synonym 1-2-fluoro-4-nitrophenyl homopiperazine, 1-2-fluoro-4-nitrophenyl-1,4-diazepane, 1-2-fluoro-4-nitrophenyl-1,4-diazaperhydroepine
PubChem CID 22291169
IUPAC Name 1-(2-fluoro-4-nitrophenyl)-1,4-diazepane
SMILES C1CNCCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])F

Specifications

Quantity 5 g
UN Number UN3259
Beilstein 10169275
Formula Weight 239.24
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2-Fluoro-4-nitrophenyl)homopiperazine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only