missing translation for 'onlineSavingsMsg'
Learn More

1,2-Ethylenediphosphonic Acid 97.0+%, TCI America™

Catalog No. E12651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12651G 1 g
E12655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12651G Supplier TCI America Supplier No. E12651G

Chemical Identifiers

CAS 6145-31-9
Molecular Formula C2H8O6P2
Molecular Weight (g/mol) 190.028
MDL Number MFCD00013958
InChI Key XYJLPCAKKYOLGU-UHFFFAOYSA-N
Synonym 1,2-ethylenediphosphonic acid, 1,2-ethanediphosphonic acid, phosphonic acid, ethylenedi, phosphonic acid, 1,2-ethanediylbis, 1,2-ethanebisphosphonic acid, 1,2-ethanediylbisphosphonic acid, ethylenediphosphonic acid, phosphonic acid, ethylenedi-6ci,7ci,8ci, ethane-1,2-diylbis phosphonicacid, ethylenebisphosphonic acid
PubChem CID 80247
IUPAC Name 2-phosphonoethylphosphonic acid
SMILES C(CP(=O)(O)O)P(=O)(O)O

Specifications

Melting Point 218°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 190.03
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Ethylenediphosphonic Acid