missing translation for 'onlineSavingsMsg'
Learn More

1,2-Ethylenediphosphonic acid, 97%

Catalog No. AAL0582814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0582814 25 g
AAL0582806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0582814 Supplier Thermo Scientific Chemicals Supplier No. L0582814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6145-31-9 | C2H8O6P2 | 190.028 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6145-31-9
Molecular Formula C2H8O6P2
Molecular Weight (g/mol) 190.028
MDL Number MFCD00013958
InChI Key XYJLPCAKKYOLGU-UHFFFAOYSA-N
Synonym 1,2-ethylenediphosphonic acid, 1,2-ethanediphosphonic acid, phosphonic acid, ethylenedi, phosphonic acid, 1,2-ethanediylbis, 1,2-ethanebisphosphonic acid, 1,2-ethanediylbisphosphonic acid, ethylenediphosphonic acid, phosphonic acid, ethylenedi-6ci,7ci,8ci, ethane-1,2-diylbis phosphonicacid, ethylenebisphosphonic acid
PubChem CID 80247
IUPAC Name 2-phosphonoethylphosphonic acid
SMILES C(CP(=O)(O)O)P(=O)(O)O

Specifications

Melting Point 217°C to 221°C
Quantity 25 g
UN Number UN3261
Beilstein 1780250
Formula Weight 190.03
Percent Purity 97%
Chemical Name or Material 1,2-Ethylenediphosphonic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECSNumber SZ7903000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only