missing translation for 'onlineSavingsMsg'
Learn More

1,2-Ethanedithiol, 98+%

Catalog No. AAL1286514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1286514 25 g
AAL1286522 100 g
AAL1286536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1286514 Supplier Thermo Scientific Chemicals Supplier No. L1286514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 540-63-6 | C2H6S2 | 94.19 g/mol

1,2-Ethanedithiol is used as a reagent in organic synthesis to convert carbonyl compounds to thioacetals (1,3-dithiolanes). It replaces the toxic reagent arsenic tirchloride, which is involved in the synthesis of membrane-permeant fluorogenic biarsenicals from precursor dyes fluorescein and resorufin. It acts bidendate ligand to form complexes with metal ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Ethanedithiol is used as a reagent in organic synthesis to convert carbonyl compounds to thioacetals (1,3-dithiolanes). It replaces the toxic reagent arsenic tirchloride, which is involved in the synthesis of membrane-permeant fluorogenic biarsenicals from precursor dyes fluorescein and resorufin. It acts bidendate ligand to form complexes with metal ions.

Solubility
Miscible with ethanol, acetone, ether and benzene. Slightly miscible with water.

Notes
Air sensitive. Keep away from sources of ignition. Incompatible with alkali metals. Avoid inhalation of its vapor.

Chemical Identifiers

CAS 540-63-6
Molecular Formula C2H6S2
Molecular Weight (g/mol) 94.19
MDL Number MFCD00004892
InChI Key VYMPLPIFKRHAAC-UHFFFAOYSA-N
Synonym 1,2-ethanedithiol, 1,2-dimercaptoethane, dithioglycol, ethylene mercaptan, ethylenedithiol, 1,2-ethanethiol, ethylenedimercaptan, dithioethyleneglycol, ethylene dithioglycol, s-ethylene dimercaptan
PubChem CID 10902
IUPAC Name ethane-1,2-dithiol
SMILES C(CS)S

Specifications

Melting Point -41°C
Density 1.123
Boiling Point 144°C to 146°C
Flash Point 45°C (113°F)
Odor Stench
Refractive Index 1.558
Linear Formula HSCH2CH2SH
Quantity 25 g
UN Number UN3071
Beilstein 505946
Sensitivity Air Sensitive
Merck Index 14,3725
Solubility Information Miscible with ethanol,acetone,ether and benzene. Slightly miscible with water.
Formula Weight 94.2
Percent Purity ≥98%
Chemical Name or Material 1,2-Ethanedithiol
Show More Show Less
Hazard Category H226-H301-H304-H312-H330
Hazard Statement GHS H Statement
H301-H330-H226-H312-H319
Toxic if swallowed.
Fatal if inhaled.
Flammable liquid and vapor.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P312-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCAPTANS, LIQUID, TOXIC, FLAMMABLE, N.O.S.
EINECSNumber 208-752-3
RTECSNumber KI3325000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only