missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxypentane 98.0+%, TCI America™

Catalog No. p-7129169
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08225ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E08225ML Supplier TCI America Supplier No. E08225ML
Only null left

Chemical Identifiers

CAS 1003-14-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00192124
InChI Key SYURNNNQIFDVCA-UHFFFAOYSA-N
Synonym 1,2-Pentylene Oxide, Propyloxirane
PubChem CID 13853
IUPAC Name 2-propyloxirane
SMILES CCCC1CO1

Specifications

Color Colorless
Boiling Point 90°C
Quantity 5 mL
UN Number 1993
Formula Weight 86.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxypentane
RTECSNumber SA2700000
TSCA Yes