Learn More
1,2-Epoxyoctane, 97%
CAS: 2984-50-1 | C8H16O | 128.22 g/mol
Supplier: Thermo Scientific Chemicals B2146322
Description
It is employed as an intermediate for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as an intermediate for organic synthesis.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
| 1,2-Epoxyoctane | |
| 0.838 | |
| 37°C (98°F) | |
| 1.42 | |
| 100 g | |
| 79912 | |
| 1,2-epoxyoctane, hexyloxirane, 1-octene oxide, octylene epoxide, oxirane, hexyl, 1-octene epoxide, octane 1,2-oxide, octene-1,2-oxide, 1,2-epoxy-n-octane, n-octene-1,2-oxide | |
| NJWSNNWLBMSXQR-UHFFFAOYNA-N | |
| 2-hexyloxirane | |
| 18126 | |
| 97% |
| 2984-50-1 | |
| 167°C | |
| C8H16O | |
| MFCD00005157 | |
| UN1993 | |
| Moisture sensitive | |
| Slightly soluble in water. | |
| CCCCCCC1CO1 | |
| 128.22 | |
| 128.22 |
Chemical Identifiers
| 2984-50-1 | |
| 128.22 | |
| NJWSNNWLBMSXQR-UHFFFAOYNA-N | |
| 18126 | |
| CCCCCCC1CO1 |
| C8H16O | |
| MFCD00005157 | |
| 1,2-epoxyoctane, hexyloxirane, 1-octene oxide, octylene epoxide, oxirane, hexyl, 1-octene epoxide, octane 1,2-oxide, octene-1,2-oxide, 1,2-epoxy-n-octane, n-octene-1,2-oxide | |
| 2-hexyloxirane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 221-047-5
RTECSNumber : RG9625000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only