missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxyoctane, 97%

Catalog No. AAB2146314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2146314 25 g
AAB2146322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2146314 Supplier Thermo Scientific Chemicals Supplier No. B2146314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2984-50-1 | C8H16O | 128.22 g/mol

It is employed as an intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate for organic synthesis.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 2984-50-1
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00005157
InChI Key NJWSNNWLBMSXQR-UHFFFAOYNA-N
Synonym 1,2-epoxyoctane, hexyloxirane, 1-octene oxide, octylene epoxide, oxirane, hexyl, 1-octene epoxide, octane 1,2-oxide, octene-1,2-oxide, 1,2-epoxy-n-octane, n-octene-1,2-oxide
PubChem CID 18126
IUPAC Name 2-hexyloxirane
SMILES CCCCCCC1CO1

Specifications

Density 0.838
Boiling Point 167°C
Flash Point 37°C (98°F)
Refractive Index 1.42
Quantity 25 g
UN Number UN1993
Beilstein 79912
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material 1,2-Epoxyoctane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 221-047-5
RTECSNumber RG9625000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only