missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxyoctadecane, tech. 85%

Catalog No. AAL0762222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0762222 100 g
AAL0762214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0762222 Supplier Thermo Scientific Chemicals Supplier No. L0762222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7390-81-0 | C18H36O | 268.485 g/mol

1,2-Epoxyoctadecane can be used to produce octadecane-1,2-diol at heating. It is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxyoctadecane can be used to produce octadecane-1,2-diol at heating. It is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Insoluble in water.

Notes
Protect from moisture. Store away from incompatible materials such as oxidizing agents. Moisture-sensitive. Incompatible with strong acids, peroxides, caustics.

Chemical Identifiers

CAS 7390-81-0
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.485
MDL Number MFCD00005152
InChI Key QBJWYMFTMJFGOL-UHFFFAOYSA-N
Synonym 1,2-epoxyoctadecane, hexadecyloxirane, oxirane, hexadecyl, octadecane, epoxy, 1,2-octadecylene oxide, ccris 2619, oxirane, 2-hexadecyl, #, 1,2-epoxy octadecane, acmc-209osu
PubChem CID 23872
IUPAC Name 2-hexadecyloxirane
SMILES CCCCCCCCCCCCCCCCC1CO1

Specifications

Melting Point 30°C to 35°C
Boiling Point 146°C to 148°C (1 mmHg)
Flash Point 176°C (348°F)
Quantity 100 g
Beilstein 115434
Solubility Information Insoluble in water.
Formula Weight 268.49
Percent Purity 85%
Grade Technical
Chemical Name or Material 1,2-Epoxyoctadecane
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 230-977-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only