missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxyoctadecane 85.0+%, TCI America™

Catalog No. p-7128928
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E031325G 25 g
E0313400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E031325G Supplier TCI America Supplier No. E031325G
Only null left

Chemical Identifiers

CAS 7390-81-0
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.485
MDL Number MFCD00005152
InChI Key QBJWYMFTMJFGOL-UHFFFAOYSA-N
Synonym 1,2-epoxyoctadecane, hexadecyloxirane, oxirane, hexadecyl, octadecane, epoxy, 1,2-octadecylene oxide, ccris 2619, oxirane, 2-hexadecyl, #, 1,2-epoxy octadecane, acmc-209osu
PubChem CID 23872
IUPAC Name 2-hexadecyloxirane
SMILES CCCCCCCCCCCCCCCCC1CO1

Specifications

Color White-Yellow
Boiling Point 137°C
Quantity 25 g
Formula Weight 268.49
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2-Epoxyoctadecane
EINECSNumber (2)-0220
TSCA Yes