Learn More
1,2-Epoxyhexane, 97%
CAS: 1436-34-6 | C6H12O | 100.16 g/mol
$107.99 - $107.99
Chemical Identifiers
| CAS | 1436-34-6 |
|---|---|
| Molecular Formula | C6H12O |
| Molecular Weight (g/mol) | 100.16 |
| MDL Number | MFCD00005154 |
| InChI Key | WHNBDXQTMPYBAT-UHFFFAOYSA-N |
| Synonym | 1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide |
| PubChem CID | 15036 |
| IUPAC Name | 2-butyloxirane |
| SMILES | CCCCC1CO1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC335550250
|
Thermo Scientific Chemicals
335550250 |
25 mL | Glass bottle |
Each for $107.99
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1436-34-6 | |
| 100.16 | |
| WHNBDXQTMPYBAT-UHFFFAOYSA-N | |
| 15036 | |
| CCCCC1CO1 |
| C6H12O | |
| MFCD00005154 | |
| 1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide | |
| 2-butyloxirane |
Specifications
| 1436-34-6 | |
| 100.0 | |
| 0.8300g/mL | |
| 15°C | |
| 97% | |
| C6H12O | |
| MFCD00005154 | |
| 0.83 | |
| WHNBDXQTMPYBAT-UHFFFAOYSA-N | |
| 2-butyloxirane | |
| 15036 | |
| 97% | |
| 1, 2-Epoxyhexane |
| 96.0 | |
| Colorless | |
| 118°C to 120°C | |
| Authentic | |
| Glass bottle | |
| 1.4040 to 1.407 | |
| 25 mL | |
| 1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide | |
| CCCCC1CO1 | |
| 100.16 | |
| 100.16 | |
| Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 215-864-6
RUO – Research Use Only