missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxyhexane, 96%

Catalog No. AAL0760914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0760914 25 g
AAL0760922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0760914 Supplier Thermo Scientific Chemicals Supplier No. L0760914

CAS: 1436-34-6 | C6H12O | 100.161 g/mol

1,2-Epoxyhexane is used as chemical reagent, organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxyhexane is used as chemical reagent, organic intermediates, fine chemicals, pharmaceutical research and development.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 1436-34-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00005154
InChI Key WHNBDXQTMPYBAT-UHFFFAOYSA-N
Synonym 1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide
PubChem CID 15036
IUPAC Name 2-butyloxirane
SMILES CCCCC1CO1

Specifications

Density 0.832
Boiling Point 118°C to 120°C
Flash Point 15°C (59°F)
Refractive Index 1.406
Quantity 25 g
UN Number UN1993
Beilstein 102568
Solubility Information Difficult to mix.
Formula Weight 100.16
Percent Purity 96%
Chemical Name or Material 1,2-Epoxyhexane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 215-864-6
RTECSNumber MO3630000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only