missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Epoxyhexane 96.0+%, TCI America™
$111.66 - $111.66
Chemical Identifiers
| CAS | 1436-34-6 |
|---|---|
| Molecular Formula | C6H12O |
| Molecular Weight (g/mol) | 100.161 |
| MDL Number | MFCD00005154 |
| InChI Key | WHNBDXQTMPYBAT-UHFFFAOYSA-N |
| Synonym | 1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide |
| PubChem CID | 15036 |
| IUPAC Name | 2-butyloxirane |
| SMILES | CCCCC1CO1 |
Chemical Identifiers
| 1436-34-6 | |
| 100.161 | |
| WHNBDXQTMPYBAT-UHFFFAOYSA-N | |
| 15036 | |
| CCCCC1CO1 |
| C6H12O | |
| MFCD00005154 | |
| 1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide | |
| 2-butyloxirane |
Specifications
| 1436-34-6 | |
| 120°C | |
| MFCD00005154 | |
| 1993 | |
| WHNBDXQTMPYBAT-UHFFFAOYSA-N | |
| 2-butyloxirane | |
| 15036 | |
| ≥96.0% (GC) | |
| 1,2-Epoxyhexane |
| Yellow | |
| C6H12O | |
| 25 mL | |
| 1,2-epoxyhexane, butyloxirane, oxirane, butyl, 1-hexene oxide, 1-hexene epoxide, 1,2-hexene oxide, epoxy-n-hexane, hexylene oxide, hexane, 1,2-epoxy, hexene, monooxide | |
| CCCCC1CO1 | |
| 100.161 | |
| 100.16 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0220
RTECSNumber : MO3630000
TSCA : No