missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxyeicosane 80.0+%, TCI America™

Catalog No. E03115G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E03115G Supplier TCI America Supplier No. E03115G
Only null left

Chemical Identifiers

CAS 19780-16-6
Molecular Formula C20H40O
Molecular Weight (g/mol) 296.54
MDL Number MFCD00059157
InChI Key BHZBVWCLMYQFQX-UHFFFAOYNA-N
Synonym 1,2-Eicosylene Oxide
PubChem CID 89221
IUPAC Name 2-octadecyloxirane
SMILES CCCCCCCCCCCCCCCCCCC1CO1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 296.54
Percent Purity ≥80.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2-Epoxyeicosane
EINECSNumber (2)-0220
TSCA No