missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxycyclopentane 97.0+%, TCI America™

Catalog No. p-7129041
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05135G 5 g
E051325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05135G Supplier TCI America Supplier No. E05135G
Only null left

Chemical Identifiers

CAS 285-67-6
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00005161
InChI Key GJEZBVHHZQAEDB-UHFFFAOYNA-N
Synonym cyclopentene oxide, 6-oxabicyclo 3.1.0 hexane, 1,2-epoxycyclopentane, cyclopentane oxide, cyclopentene epoxide, epoxycyclopentane, cis-1,2-epoxycyclopentane, cyclopentane, 1,2-epoxy, 6-oxa-bicyclo 3.1.0 hexane
PubChem CID 9244
IUPAC Name 6-oxabicyclo[3.1.0]hexane
SMILES C1CC2OC2C1

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
UN Number 1993
Formula Weight 84.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxycyclopentane
RTECSNumber RN8935000
TSCA Yes