missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxycyclohexane 98.0+%, TCI America™

Catalog No. p-7134170
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O007125ML 25 mL
O0071100ML 100 mL
O0071500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O007125ML Supplier TCI America Supplier No. O007125ML
Only null left

Chemical Identifiers

CAS 286-20-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00005162
InChI Key ZWAJLVLEBYIOTI-UHFFFAOYSA-N
Synonym cyclohexene oxide, 7-oxabicyclo 4.1.0 heptane, 1,2-epoxycyclohexane, cyclohexene epoxide, cyclohexylene oxide, tetramethyleneoxirane, cyclohexene 1-oxide, epoxycyclohexane, cyclohexeneoxide, 1,2-cyclohexene oxide
PubChem CID 9246
IUPAC Name 7-oxabicyclo[4.1.0]heptane
SMILES C1CCC2C(C1)O2

Specifications

Color Colorless
Boiling Point 130°C
Quantity 25 mL
UN Number 1993
Formula Weight 98.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxycyclohexane
EINECSNumber (3)-3263
RTECSNumber RN7175000
TSCA Yes