missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-9-decene 96.0+%, TCI America™

Catalog No. e04115ml
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04115ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04115ML Supplier TCI America Supplier No. E04115ML
Only null left

Specifications

CAS 85721-25-1
Color Colorless
Boiling Point 80°C
Molecular Formula C10H18O
Quantity 5 mL
MDL Number MFCD00069427
Synonym 1,2-epoxy-9-decene, oxirane, 7-octenyl, oct-7-en-1-yloxirane, 2-7-octen-1-yl oxirane, 2-oct-7-en-1-yl oxirane, oct-7-enyloxirane, 7-octenyl oxirane, zlchem 822, acmc-20apkm, 2-7-octenyl oxirane
InChI Key FCZHJHKCOZGQJZ-UHFFFAOYSA-N
SMILES C=CCCCCCCC1CO1
IUPAC Name 2-oct-7-enyloxirane
Molecular Weight (g/mol) 154.253
PubChem CID 543758
Formula Weight 154.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxy-9-decene
Show More Show Less