missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) 98.0+%, TCI America™

Catalog No. p-7129246
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E095625ML 25 mL
E0956250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E095625ML Supplier TCI America Supplier No. E095625ML
Only null left

Chemical Identifiers

CAS 106-86-5
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00022356
InChI Key SLJFKNONPLNAPF-UHFFFAOYSA-N
Synonym 4-Vinyl-1,2-epoxycyclohexane, 3-Vinyl-7-oxabicyclo[4.1.0]heptane
PubChem CID 7832
IUPAC Name 4-ethenyl-7-oxabicyclo[4.1.0]heptane
SMILES C=CC1CCC2C(C1)O2

Specifications

Melting Point -100°C
Color Colorless
Boiling Point 159°C
Quantity 25 mL
UN Number 3271
Formula Weight 124.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxy-4-vinylcyclohexane (mixture of isomers)
EINECSNumber (3)-2296
RTECSNumber RN8770000
TSCA Yes
Recommended Storage Refrigerator