missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-3-methylbutane, 98+%

CAS: 1438-14-8 | C5H10O | 86.134 g/mol

Supplier:  Thermo Scientific Chemicals L1929103

 View more versions of this product

Catalog No. AAL1929103

Only null left
Explore available promotions

Description

Description

1,2-Epoxy-3-methylbutane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-3-methylbutane is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Store under inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.
Specifications

Specifications

1,2-Epoxy-3-methylbutane
0.815
−12°C (10°F)
C5H10O
MFCD02683501
UN1993
Moisture sensitive
Not miscible or difficult to mix in water.
CC(C)C1CO1
86.134
86.13
1438-14-8
72°C to 75°C
Recognizable
1.3915
1 g
102418
2-isopropyloxirane, 1,2-epoxy-3-methylbutane, isopropyloxirane, oxirane, 1-methylethyl, butane, 1,2-epoxy-3-methyl, 2-propan-2-yl oxirane, oxirane,2-1-methylethyl, 1-methylethyl-oxirane, isopropylethylene oxide, acmc-1asns
REYZXWIIUPKFTI-UHFFFAOYSA-N
2-propan-2-yloxirane
102618
≥98%

Chemical Identifiers

1438-14-8
86.134
REYZXWIIUPKFTI-UHFFFAOYSA-N
102618
CC(C)C1CO1
C5H10O
MFCD02683501
2-isopropyloxirane, 1,2-epoxy-3-methylbutane, isopropyloxirane, oxirane, 1-methylethyl, butane, 1,2-epoxy-3-methyl, 2-propan-2-yl oxirane, oxirane,2-1-methylethyl, 1-methylethyl-oxirane, isopropylethylene oxide, acmc-1asns
2-propan-2-yloxirane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 215-869-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only