missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-3-methylbutane, 98+%

Catalog No. AAL1929106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1929106 5 g
AAL1929103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1929106 Supplier Thermo Scientific Chemicals Supplier No. L1929106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 1438-14-8 | C5H10O | 86.134 g/mol

1,2-Epoxy-3-methylbutane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-3-methylbutane is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Store under inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.

Chemical Identifiers

CAS 1438-14-8
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD02683501
InChI Key REYZXWIIUPKFTI-UHFFFAOYSA-N
Synonym 2-isopropyloxirane, 1,2-epoxy-3-methylbutane, isopropyloxirane, oxirane, 1-methylethyl, butane, 1,2-epoxy-3-methyl, 2-propan-2-yl oxirane, oxirane,2-1-methylethyl, 1-methylethyl-oxirane, isopropylethylene oxide, acmc-1asns
PubChem CID 102618
IUPAC Name 2-propan-2-yloxirane
SMILES CC(C)C1CO1

Specifications

Density 0.815
Boiling Point 72°C to 75°C
Flash Point −12°C (10°F)
Odor Recognizable
Refractive Index 1.3915
Quantity 5 g
UN Number UN1993
Beilstein 102418
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 86.13
Percent Purity ≥98%
Chemical Name or Material 1,2-Epoxy-3-methylbutane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 215-869-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only