missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-3,3,3-trifluoropropane, 98%

Catalog No. AAL0986903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0986903 1 g
AAL0986906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0986903 Supplier Thermo Scientific Chemicals Supplier No. L0986903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 359-41-1 | C3H3F3O | 112.051 g/mol

1,2-Epoxy-3,3,3-trifluoropropane is used for the synthesis of trifluorolactic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-3,3,3-trifluoropropane is used for the synthesis of trifluorolactic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible with water.

Notes
Moisture Sensitive. Protect from humidity and water. Incompatible materials are heat, oxidizing agents and water. Store away from moisture.

Chemical Identifiers

CAS 359-41-1
Molecular Formula C3H3F3O
Molecular Weight (g/mol) 112.051
MDL Number MFCD00041506
InChI Key AQZRARFZZMGLHL-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-2,3-epoxypropane, 2-trifluoromethyl oxirane, 3,3,3-trifluoro-1,2-epoxypropane, 1,2-epoxy-3,3,3-trifluoropropane, oxirane, trifluoromethyl, 3,3,3-trifluoropropane epoxide, pubchem20840, trifluoromethyl oxirane, acmc-20apk3
PubChem CID 520769
IUPAC Name 2-(trifluoromethyl)oxirane
SMILES C1C(O1)C(F)(F)F

Specifications

Density 1.3
Boiling Point 38°C to 40°C
Flash Point −40°C (−40°F)
Refractive Index 1.297
Quantity 1 g
UN Number UN1992
Beilstein 103730
Sensitivity Moisture sensitive
Solubility Information Not miscible with water.
Formula Weight 112.05
Percent Purity 98%
Chemical Name or Material 1,2-Epoxy-3,3,3-trifluoropropane
Show More Show Less
Hazard Category H225-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H225-H301-H311-H331-H315-H319-H335
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only