missing translation for 'onlineSavingsMsg'
Learn More

1,2-Distearoyl-sn-glycero-3-phosphocholine 95.0+%, TCI America™

Catalog No. D3926200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3926200MG 200 mg
D39261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3926200MG Supplier TCI America Supplier No. D3926200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 816-94-4
Molecular Formula C44H88NO8P
Molecular Weight (g/mol) 790.16
MDL Number MFCD00036905
InChI Key NRJAVPSFFCBXDT-HUESYALOSA-N
Synonym 1,2-Dioctadecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(stearoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, DSPC
PubChem CID 94190
ChEBI CHEBI:83718
IUPAC Name (2-{[(2R)-2,3-bis(octadecanoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
SMILES CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCCCC

Specifications

Melting Point 236°C
Color White
Quantity 200 mg
Formula Weight 790.16
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Distearoyl-sn-glycero-3-phosphocholine
TSCA No
Recommended Storage Freezer