missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1,2-Diphenylethylenediamine 98.0+%, TCI America™

Catalog No. D25195G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25195G Supplier TCI America Supplier No. D25195G
Only null left

Chemical Identifiers

CAS 16635-95-3
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.30
MDL Number MFCD00709169
InChI Key PONXTPCRRASWKW-UHFFFAOYNA-N
Synonym (+/-)-1,2-Diamino-1,2-diphenylethane
PubChem CID 6931238
IUPAC Name 1,2-diphenylethane-1,2-diamine
SMILES NC(C(N)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 81°C
Color White-Yellow
Quantity 5 g
Formula Weight 212.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-1,2-Diphenylethylenediamine
TSCA No
Recommended Storage Refrigerator