missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diphenylethylamine 97.0+%, TCI America™

Catalog No. p-7126613
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11445G 5 g
D114425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D11445G Supplier TCI America Supplier No. D11445G
Only null left

Chemical Identifiers

CAS 25611-78-3
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD00008063
InChI Key DTGGNTMERRTPLR-UHFFFAOYSA-N
Synonym 1-Amino-1,2-diphenylethane, alpha-Benzylbenzylamine
PubChem CID 26482
IUPAC Name 1,2-diphenylethanamine
SMILES C1=CC=C(C=C1)CC(C2=CC=CC=C2)N

Specifications

Color Yellow
Quantity 5 g
Formula Weight 197.28
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,2-Diphenylethylamine
RTECSNumber SI2299000
TSCA No