missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diphenylethane, 98+%

Catalog No. AAA1461736
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1461736 500 g
AAA1461714 25 g
AAA1461722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1461736 Supplier Thermo Scientific Chemicals Supplier No. A1461736
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103-29-7 | C14H14 | 182.27 g/mol

1,2-Diphenylethane is used as solvent for nitro fibre and in synthesis of other organic chemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diphenylethane is used as solvent for nitro fibre and in synthesis of other organic chemical products.

Solubility
Soluble in ether, chloroform

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 103-29-7
Molecular Formula C14H14
Molecular Weight (g/mol) 182.27
MDL Number MFCD00004796
InChI Key QWUWMCYKGHVNAV-UHFFFAOYSA-N
Synonym 1,2-diphenylethane, bibenzyl, dibenzyl, s-diphenylethane, 2-phenylethyl benzene, 1,2-dihydrostilbene, dibenzil, sym-diphenylethane, dihydrostilbene, benzene, 1,1'-1,2-ethanediyl bis
PubChem CID 7647
ChEBI CHEBI:34047
IUPAC Name 2-phenylethylbenzene
SMILES C(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 50°C to 53°C
Density 1.013
Boiling Point 284°C
Flash Point >110°C (230°F)
Quantity 500 g
Beilstein 508068
Merck Index 14,1198
Solubility Information Soluble in ether, chloroform
Formula Weight 182.27
Percent Purity ≥98%
Chemical Name or Material 1,2-Diphenylethane
Show More Show Less
EINECSNumber 203-096-4
RTECSNumber DT4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only