missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diphenylcyclopropane, cis + trans, 97%

Catalog No. p-7044947
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0883304 2 g
AAL0883309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0883304 Supplier Thermo Scientific Chemicals Supplier No. L0883304
Only null left

CAS: 29881-14-9 | C15H14 | 194.277 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29881-14-9
Molecular Formula C15H14
Molecular Weight (g/mol) 194.277
MDL Number MFCD00040875
InChI Key ZSIYTDQNAOYUNE-UHFFFAOYSA-N
Synonym 1,2-diphenylcyclopropane, 2-phenylcyclopropyl benzene, 1,2-diphenylcyclopropane, cis + trans, benzene, 1,1'-1,2-cyclopropanediyl bis, 1,2-diphenylcyclopropane, trans, acmc-1cckp, trans-1,1'-1,2-cyclopropanediyl bisbenzene, benzene, 1,1'-1,2-cyclopropanediyl bis-, cis, 2-phenylcyclopropyl benzene #, benzene,1,1'-1,2-cyclopropanediyl bis
PubChem CID 70824
IUPAC Name (2-phenylcyclopropyl)benzene
SMILES C1C(C1C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Density 1.02
Boiling Point 175°C to 177°C (14 mmHg)
Refractive Index 1.595
Quantity 2 g
Beilstein 2043299
Formula Weight 194.28
Percent Purity 97%
Chemical Name or Material 1,2-Diphenylcyclopropane, Cis + Trans
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only