missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dimyristoyl-sn-glycero-3-phosphocholine 97.0+%, TCI America™
$210.63 - $598.37
Chemical Identifiers
| CAS | 18194-24-6 |
|---|---|
| Molecular Formula | C36H72NO8P |
| Molecular Weight (g/mol) | 677.945 |
| MDL Number | MFCD00036901 |
| InChI Key | CITHEXJVPOWHKC-UUWRZZSWSA-N |
| Synonym | 1,2-Ditetradecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(tetradecanoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, DMPC |
| PubChem CID | 5459377 |
| ChEBI | CHEBI:45240 |
| IUPAC Name | [(2R)-2,3-di(tetradecanoyloxy)propyl] 2-(trimethylazaniumyl)ethyl phosphate |
| SMILES | CCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D3924200MG
|
TCI America
D3924200MG |
200 mg |
Each for $210.63
|
|
|||||
|
D39241G
|
TCI America
D39241G |
1 g |
Each for $598.37
|
|
|||||
Chemical Identifiers
| 18194-24-6 | |
| 677.945 | |
| CITHEXJVPOWHKC-UUWRZZSWSA-N | |
| 5459377 | |
| [(2R)-2,3-di(tetradecanoyloxy)propyl] 2-(trimethylazaniumyl)ethyl phosphate |
| C36H72NO8P | |
| MFCD00036901 | |
| 1,2-Ditetradecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(tetradecanoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, DMPC | |
| CHEBI:45240 | |
| CCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCC |
Specifications
| 18194-24-6 | |
| White | |
| MFCD00036901 | |
| 1,2-Ditetradecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(tetradecanoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, DMPC | |
| CCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCC | |
| 677.945 | |
| CHEBI:45240 | |
| ≥97.0% (HPLC,T) | |
| 1,2-Dimyristoyl-sn-glycero-3-phosphocholine |
| 220°C | |
| C36H72NO8P | |
| 200 mg | |
| CITHEXJVPOWHKC-UUWRZZSWSA-N | |
| [(2R)-2,3-di(tetradecanoyloxy)propyl] 2-(trimethylazaniumyl)ethyl phosphate | |
| 5459377 | |
| 677.94 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer