missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dimyristoyl-sn-glycero-3-phosphocholine 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 220°C |
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 677.94 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,2-Dimyristoyl-sn-glycero-3-phosphocholine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |