missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethylimidazole 98.0+%, TCI America™

Catalog No. D1562500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1562500G 500 g
D156225G 25 g
D1562100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1562500G Supplier TCI America Supplier No. D1562500G

Chemical Identifiers

CAS 1739-84-0
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005294
InChI Key GIWQSPITLQVMSG-UHFFFAOYSA-N
Synonym 1,2-dimethyl-1h-imidazole, 1h-imidazole, 1,2-dimethyl, imidazole, 1,2-dimethyl, 1h-imidazole, dimethyl, unii-q41bc3grjb, q41bc3grjb, lupragen r dmi, 1,2-dimethylimdazole, 1,2 dimethylimidazole, n,2-dimethylimidazole
PubChem CID 15617
IUPAC Name 1,2-dimethyl-1H-imidazole
SMILES CN1C=CN=C1C

Specifications

Color White-Yellow
Boiling Point 205°C
Quantity 500 g
UN Number 3263
Formula Weight 96.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2-Dimethylimidazole