missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethylimidazole, 98%

Catalog No. AAB2354414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2354414 25 g
AAB2354422 100 g
AAB2354436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2354414 Supplier Thermo Scientific Chemicals Supplier No. B2354414

CAS: 1739-84-0 | C5H8N2 | 96.13 g/mol

1,2-Dimethylimidazole is used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It also can be used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dimethylimidazole is used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It also can be used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.

Solubility
Soluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 1739-84-0
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005294
InChI Key GIWQSPITLQVMSG-UHFFFAOYSA-N
Synonym 1,2-dimethyl-1h-imidazole, 1h-imidazole, 1,2-dimethyl, imidazole, 1,2-dimethyl, 1h-imidazole, dimethyl, unii-q41bc3grjb, q41bc3grjb, lupragen r dmi, 1,2-dimethylimdazole, 1,2 dimethylimidazole, n,2-dimethylimidazole
PubChem CID 15617
IUPAC Name 1,2-dimethylimidazole
SMILES CN1C=CN=C1C

Specifications

Melting Point 35°C to 38°C
Density 1.084
Boiling Point 203°C to 205°C
Flash Point 92°C (197°F)
Quantity 25 g
Beilstein 108457
Solubility Information Soluble in water.
Formula Weight 96.13
Percent Purity 98%
Chemical Name or Material 1,2-Dimethylimidazole
Hazard Category H227-H302-H315-H317-H318-H335
Hazard Statement GHS H Statement
H318-H302-H315
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement P210-P234-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P390-P501c
EINECSNumber 217-101-2
RTECSNumber NI4838854
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only