missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Dimethylaminoethyl)-4-methylpiperazine 98.0+%, TCI America™
$59.65 - $186.41
Chemical Identifiers
| CAS | 104-19-8 |
|---|---|
| Molecular Formula | C9H21N3 |
| Molecular Weight (g/mol) | 171.288 |
| MDL Number | MFCD00059773 |
| InChI Key | XFLSMWXCZBIXLV-UHFFFAOYSA-N |
| PubChem CID | 66908 |
| IUPAC Name | N,N-dimethyl-2-(4-methylpiperazin-1-yl)ethanamine |
| SMILES | CN1CCN(CC1)CCN(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D176625ML
|
TCI America
D176625ML |
25 mL |
Each for $59.65
|
|
|||||
|
D1766500ML
|
TCI America
D1766500ML |
500 mL |
Each for $186.41
|
|
|||||
Chemical Identifiers
| 104-19-8 | |
| 171.288 | |
| XFLSMWXCZBIXLV-UHFFFAOYSA-N | |
| N,N-dimethyl-2-(4-methylpiperazin-1-yl)ethanamine |
| C9H21N3 | |
| MFCD00059773 | |
| 66908 | |
| CN1CCN(CC1)CCN(C)C |
Specifications
| 104-19-8 | |
| 96°C | |
| MFCD00059773 | |
| 2922 | |
| CN1CCN(CC1)CCN(C)C | |
| 171.288 | |
| 171.29 | |
| Liquid |
| Yellow | |
| C9H21N3 | |
| 25 mL | |
| XFLSMWXCZBIXLV-UHFFFAOYSA-N | |
| N,N-dimethyl-2-(4-methylpiperazin-1-yl)ethanamine | |
| 66908 | |
| ≥98.0% (GC,T) | |
| 1-(2-Dimethylaminoethyl)-4-methylpiperazine |
Safety and Handling
EINECSNumber : (5)-0964
RTECSNumber : TL6125000
TSCA : Yes