missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Dimethylaminoethyl)-4-methylpiperazine 98.0+%, TCI America™

Catalog No. D1766500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1766500ML 500 mL
D176625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1766500ML Supplier TCI America Supplier No. D1766500ML

Chemical Identifiers

CAS 104-19-8
Molecular Formula C9H21N3
Molecular Weight (g/mol) 171.288
MDL Number MFCD00059773
InChI Key XFLSMWXCZBIXLV-UHFFFAOYSA-N
PubChem CID 66908
IUPAC Name N,N-dimethyl-2-(4-methylpiperazin-1-yl)ethanamine
SMILES CN1CCN(CC1)CCN(C)C

Specifications

Color Yellow
Boiling Point 96°C
Quantity 500 mL
UN Number 2922
Formula Weight 171.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(2-Dimethylaminoethyl)-4-methylpiperazine
EINECSNumber (5)-0964
RTECSNumber TL6125000
TSCA Yes