missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethyl-3-propylimidazolium Iodide 98.0+%, TCI America™

Supplier:  TCI America D390325G

 View more versions of this product

Catalog No. D390325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,2-Dimethyl-3-propylimidazolium Iodide
White-Yellow
25 g
CCCN1C=C[N+](=C1C)C.[I-]
266.126
266.13
Crystalline Powder
218151-78-1
C8H15IN2
ISHFYECQSXFODS-UHFFFAOYSA-M
1,2-dimethyl-3-propylimidazol-1-ium;iodide
11459786
≥98.0% (HPLC,T)

Chemical Identifiers

218151-78-1
266.126
11459786
CCCN1C=C[N+](=C1C)C.[I-]
C8H15IN2
ISHFYECQSXFODS-UHFFFAOYSA-M
1,2-dimethyl-3-propylimidazol-1-ium;iodide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions