missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethyl-3-propylimidazolium Iodide 98.0+%, TCI America™

Catalog No. D39035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39035G 5 g
D390325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39035G Supplier TCI America Supplier No. D39035G

Chemical Identifiers

CAS 218151-78-1
Molecular Formula C8H15IN2
Molecular Weight (g/mol) 266.126
InChI Key ISHFYECQSXFODS-UHFFFAOYSA-M
PubChem CID 11459786
IUPAC Name 1,2-dimethyl-3-propylimidazol-1-ium;iodide
SMILES CCCN1C=C[N+](=C1C)C.[I-]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 266.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dimethyl-3-propylimidazolium Iodide