missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dimethyl-1,1,2,2-tetraphenyldisilane 98.0+%, TCI America™
Supplier: TCI America D50315G
Specifications
| 1,2-Dimethyl-1,1,2,2-tetraphenyldisilane | |
| 144°C | |
| 185°C | |
| MFCD00051536 | |
| JNZRJYXUMDPPRK-UHFFFAOYSA-N | |
| methyl-[methyl(diphenyl)silyl]-diphenylsilane | |
| 70895 | |
| ≥98.0% (GC) |
| 1172-76-5 | |
| White | |
| C26H26Si2 | |
| 5 g | |
| C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)[Si](C)(C3=CC=CC=C3)C4=CC=CC=C4 | |
| 394.664 | |
| 394.66 | |
| Crystalline Powder |
Chemical Identifiers
| 1172-76-5 | |
| 394.664 | |
| JNZRJYXUMDPPRK-UHFFFAOYSA-N | |
| methyl-[methyl(diphenyl)silyl]-diphenylsilane |
| C26H26Si2 | |
| MFCD00051536 | |
| 70895 | |
| C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)[Si](C)(C3=CC=CC=C3)C4=CC=CC=C4 |
Safety and Handling
TSCA : Yes