missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxypropane 98.0+%, TCI America™

Catalog No. d428125ml
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D428125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D428125ML Supplier TCI America Supplier No. D428125ML
Only null left

Chemical Identifiers

CAS 7778-85-0
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00010331
InChI Key LEEANUDEDHYDTG-UHFFFAOYNA-N
Synonym Propylene Glycol Dimethyl Ether
PubChem CID 24509
IUPAC Name 1,2-dimethoxypropane
SMILES COCC(C)OC

Specifications

Color Colorless
Boiling Point 96°C
Quantity 25 mL
UN Number 3271
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dimethoxypropane
EINECSNumber (7)-1321
RTECSNumber TY1145000
TSCA No